Learn More
Invitrogen™ CD8/CD28 Antibody Cocktail, FITC, PE
Description
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD28 (clone B-23, isotype IgG1).
Specifications
Specifications
| Antigen | CD8/CD28 |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Clone | 17D8, B-23, B-23 |
| Conjugate | FITC/PE |
| Formulation | PBS with 0.2% carrier protein and 0.08% sodium azide |
| Gene | CD28 |
| Gene Accession No. | P01732, P10747, P10966 |
| Gene Alias | CD28; CD28 antigen; CD28 molecule; CD8; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8b; CD8b molecule; CD8B1; CD8beta; LEU2; Leu-2; Leu2 T-lymphocyte antigen; LY3; LYT3; MAL; MGC138290; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell-specific surface glycoprotein CD28; T-lymphocyte differentiation antigen T8/Leu-2; Tp44 |
| Gene Symbols | CD28, CD8A, CD8B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.