Learn More
CD8 Mouse anti-Human, PE, Clone: HIT8A, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555635
Description
Reacts with the α subunit (32kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 are not cross-blocking.
Host Species: Mouse
Clone: HIT8a
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD8 | |
Monoclonal | |
PE | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
HIT8A | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.