Learn More
CD8 Mouse anti-Human, PE, Clone: HIT8A, BD
Mouse Monoclonal Antibody
$74.00 - $158.70
Specifications
Antigen | CD8 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Host Species | Mouse |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560959
|
BD Biosciences
560959 |
25 Tests |
Each for $74.00
|
|
|||||
BDB555635
|
BD Biosciences
555635 |
100 Tests |
Each for $158.70
|
|
|||||
Description
Reacts with the α subunit (32 kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 (Cat. No. 555364) are not cross-blocking.
Specifications
CD8 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.