Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8α Rat anti-Mouse, FITC, Clone: 53-6.7, Southern Biotech™

Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 155002S
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD8α | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 | |
Spleen cells or thymocyte membranes | |
RUO | |
Mouse | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
53-6.7 | |
FITC | |
Rat | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction