Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8α Rat anti-Mouse, FITC, Clone: 53-6.7, Southern Biotech™

Rat Monoclonal Antibody
$124.10 - $280.00
Specifications
Antigen | CD8α |
---|---|
Clone | 53-6.7 |
Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
OB1550-02S
![]() |
Southern Biotechnology Associates
155002S |
0.1 mg |
Each for $124.10
|
|
|||||
OB1550-02
![]() |
Southern Biotechnology Associates
155002 |
0.5 mg |
Each for $280.00
|
|
|||||
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD8α | |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
FITC | |
RUO | |
Supplied in PBS/NaN3 | |
IgG2a | |
Store at 2-8°C; Protect from light |
53-6.7 | |
Monoclonal | |
Rat | |
Mouse | |
Spleen cells or thymocyte membranes | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title