Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Antibody (1.3.3.22), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB154639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154639 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB154639 Supplier Novus Biologicals Supplier No. NBP312080DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD81 Monoclonal antibody specifically detects CD81 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen CD81
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 1.3.3.22
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with a B-Cell line derived from a Burkitt lymphoma.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 975
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.