Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Antibody (1D6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB015969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015969 Supplier Novus Biologicals Supplier No. NBP281096APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD81 Monoclonal specifically detects CD81 in Human, Goat, Chimpanzee, Sheep samples. It is validated for Flow Cytometry.

Specifications

Antigen CD81
Applications Flow Cytometry
Classification Monoclonal
Clone 1D6
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Gene Symbols CD81
Host Species Rabbit
Immunogen This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 975
Test Specificity This antibody is specific for human CD81, which is expressed on most tissues with the exception of red blood cells and platelets.
Target Species Human, Goat, Chimpanzee, Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.