Learn More
Description
Specifications
Specifications
| Antigen | CD81 |
| Applications | CyTOF, Cytometric Bead Assay Standard, Dot Blot, ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 1D6 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 0.2 - 1 ug per million cells, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, CyTOF-ready |
| Gene Accession No. | P60033 |
| Gene Alias | CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 |
| Gene Symbols | CD81 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
