Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Antibody (2F7) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP128136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128136 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128136 Supplier Novus Biologicals Supplier No. NBP128136
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD81 Monoclonal specifically detects CD81 in Human, Mouse, Hamster samples. It is validated for Flow Cytometry.

Specifications

Antigen CD81
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, Blocking Assay
Classification Monoclonal
Clone 2F7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Gene Symbols CD81
Host Species Armenian Hamster
Immunogen Mouse epithelial cell line PAM212
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 975
Test Specificity Mouse CD81 (Mr 25 kDa), a member of the transmembrane 4 integral membrane protein family
Target Species Human, Mouse, Hamster
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.