Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Antibody (2F7), HRP, Novus Biologicals™
SDP

Catalog No. NBP128136H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128136H 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128136H Supplier Novus Biologicals Supplier No. NBP128136H
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD81 Monoclonal specifically detects CD81 in Human, Mouse, Hamster samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CD81
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2F7
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Gene Symbols CD81
Host Species Armenian Hamster
Immunogen Mouse epithelial cell line PAM212
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 975
Test Specificity Mouse CD81 (Mr 25 kDa), a member of the transmembrane 4 integral membrane protein family
Target Species Human, Mouse, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.