Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD82/Kai-1 Mouse anti-Human, Clone: TS82b, Novus Biologicals™
SDP

Catalog No. NB160255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160255 200 μg
NB160256 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB160255 Supplier Novus Biologicals Supplier No. NBP318118200UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD82/Kai-1 Monoclonal antibody specifically detects CD82/Kai-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation

Specifications

Antigen CD82/Kai-1
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone TS82b
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunoprecipitation
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9
Host Species Mouse
Immunogen Jurkat and HEL cell lines. Hybridoma: Myeloma X63/AG8. x Balb/c spleen cells
Purification Method Affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3732
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.