Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD82/Kai-1 Rabbit anti-Human, Clone: 71, Novus Biologicals™
SDP

Catalog No. NB021636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021636 50 μg
NB021635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021636 Supplier Novus Biologicals Supplier No. NBP29008650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD82/Kai-1 Monoclonal specifically detects CD82/Kai-1 in Human samples. It is validated for ELISA, Sandwich ELISA Capture.

Specifications

Antigen CD82/Kai-1
Applications ELISA
Classification Monoclonal
Clone 71
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9
Host Species Rabbit
Immunogen Recombinant Human CD82/Kai-1 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3732
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.