Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD83 Antibody (101), PE, Novus Biologicals™
SDP

Catalog No. NB383152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383152 Supplier Novus Biologicals Supplier No. NBP312771PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD83 Monoclonal antibody specifically detects CD83 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CD83
Applications Flow Cytometry
Classification Monoclonal
Clone 101
Conjugate PE
Formulation PBS
Gene Alias B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 (Uniprot#: O88324; Met1-Arg133).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9308
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.