Learn More
Description
Specifications
Specifications
| Antigen | CD83 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 104 |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CD83 (Q01151-1; Met1-Ala143). |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
