Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD83 Antibody (982502), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB314168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314168 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314168 Supplier Novus Biologicals Supplier No. FAB17741APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD83
Applications Flow Cytometry
Classification Monoclonal
Clone 982502
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human CD83, Met1-Ala143, Accession # Q01151
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9308
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.