Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD83 Antibody (MM0179-4B34) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP212180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212180 0.1 mg
NB019086 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP212180 Supplier Novus Biologicals Supplier No. NBP212180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD83 Monoclonal specifically detects CD83 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen CD83
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone MM0179-4B34
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-100, Immunohistochemistry 1:20-1:100, Immunohistochemistry-Frozen 1:20-100, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. Q01151
Gene Alias B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83
Gene Symbols CD83
Host Species Mouse
Immunogen Human recombinant CD83
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9308
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.