Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD83 Antibody (MM0179-4B34) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $584.50
Specifications
Antigen | CD83 |
---|---|
Clone | MM0179-4B34 |
Concentration | LYOPH |
Applications | Flow Cytometry, CyTOF, Immunohistochemistry (Frozen), Immunohistochemistry |
Classification | Monoclonal |
Description
CD83 Monoclonal specifically detects CD83 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
CD83 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
CD83 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0179-4B34 | |
Flow Cytometry, CyTOF, Immunohistochemistry (Frozen), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Dendritic Cell Markers | |
Q01151 | |
9308 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title