Learn More
Description
Specifications
Specifications
| Antigen | CD84/SLAMF5 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4 |
| Conjugate | Alexa Fluor 532 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CD84/SLAMF5 (NP_003865.1; Met1-Gly225). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
