Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD84/SLAMF5 Antibody (153-4D9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB121665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121665 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121665 Supplier Novus Biologicals Supplier No. NBP311578
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD84/SLAMF5 Monoclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunofluorescence

Specifications

Antigen CD84/SLAMF5
Applications Flow Cytometry, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone 153-4D9
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Gene Alias CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B
Host Species Mouse
Immunogen MLR-stimulated lymphocytes
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8832
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.