Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD84/SLAMF5 Antibody (CD84.1.21), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N106929A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106929A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106929A32 Supplier Novus Biologicals Supplier No. NB10065929AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD84/SLAMF5
Applications Flow Cytometry
Classification Monoclonal
Clone CD84.1.21
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B
Gene Symbols CD84
Host Species Mouse
Immunogen Mouse pre-B cell line 300.19 transfected with CD84 cDNA.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8832
Test Specificity NB100-65929 recognizes the human CD84 cell surface antigen, a highly glycosylated cell surface antigen of 64-82kD. CD84 is predominantly expressed by B lymphocytes, monocytes and macrophages, although lower levels of expression have also been reported on T cells, granulocytes and platelets.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.