Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD84/SLAMF5 Rabbit anti-Human, Clone: 6, Novus Biologicals™
SDP

Catalog No. NB020018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB020018 Supplier Novus Biologicals Supplier No. NBP289267100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen CD84/SLAMF5
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B
Host Species Rabbit
Immunogen Recombinant Human CD84/SLAMF5 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8832
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.