Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Antibody (1021007), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348911 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348911 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348911 Supplier Novus Biologicals Supplier No. FAB25292MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD9 Monoclonal specifically detects CD9 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen CD9
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1021007
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human CD9, Ser112-Ile195, Accession # P21926
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 928
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.