Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Antibody (EM-04), PerCP, Novus Biologicals™
SDP

Catalog No. NBP144876JP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144876JP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144876JP Supplier Novus Biologicals Supplier No. NBP144876PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD9 Monoclonal specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD9
Applications Flow Cytometry
Classification Monoclonal
Clone EM-04
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Gene Symbols CD9
Host Species Rat
Immunogen Permeabilized murine bone marrow-derived mast cells (BMMC).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Embryonic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 928
Test Specificity The rat monoclonal antibody EM-04 recognizes CD9 antigen, a 24 kDa transmembrane protein expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.