Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Antibody (HI9a), Novus Biologicals™
SDP

Catalog No. NB170641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170641 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB170641 Supplier Novus Biologicals Supplier No. NBP318795
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD9 Monoclonal antibody specifically detects CD9 in Human, Bovine, Canine, Equine, Rabbit, Sheep samples. It is validated for Flow Cytometry

Specifications

Antigen CD9
Applications Flow Cytometry
Classification Monoclonal
Clone HI9a
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 ug/10 million cells in 100 ul
Formulation PBS, pH 7.2
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Host Species Mouse
Immunogen A recombinant protein of human CD9
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 928
Target Species Human, Bovine, Canine, Equine, Rabbit, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.