Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Antibody (IVA50), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB50494A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50494A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50494A350 Supplier Novus Biologicals Supplier No. NB500494AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD9 Monoclonal antibody specifically detects CD9 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD9
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone IVA50
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Gene Symbols CD9
Host Species Mouse
Immunogen Bovine thrombocytes.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 928
Test Specificity The antibody IVA50 reacts with CD9 antigen (bovine), a 24 kDa single transmembrane polypeptide expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes.
Target Species Human, Bovine
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.