Learn More
Description
Specifications
Specifications
| Antigen | CD9 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | MM0180-5H15 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Flow Cytometry |
| Gene Accession No. | P21926 |
| Gene Alias | BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) |
| Gene Symbols | CD9 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
