Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Antibody (MM2/57), PE, Novus Biologicals™
SDP

Catalog No. NB001906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001906 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001906 Supplier Novus Biologicals Supplier No. NBP1283651000UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD9 Monoclonal specifically detects CD9 in Human, Mouse, Canine, Feline, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen CD9
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM2/57
Concentration 0.1 mg/mL
Conjugate PE
Dilution Western Blot 1:100-1:2000, Flow Cytometry 10 μL/106 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Gene Symbols CD9
Host Species Mouse
Immunogen The immunogen is a peptide made to the amino acid region CD9
Quantity 1000 μL
Gene ID (Entrez) 928
Target Species Human, Canine, Feline, Hamster, Monkey, Rabbit
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.