Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.2 Antibody (30-H12), FITC, Novus Biologicals™
SDP

Catalog No. NB001900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001900 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001900 Supplier Novus Biologicals Supplier No. NBP128037500UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD90.2 Monoclonal specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.

Specifications

Antigen CD90.2
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay, Immunocytochemistry
Classification Monoclonal
Clone 30-H12
Concentration 0.1 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1 μg/106 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Immunocytochemistry
Formulation PBS
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Gene Symbols THY1
Host Species Rat
Immunogen Mouse thymus or spleen
Quantity 500 μg
Research Discipline Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Gene ID (Entrez) 7070
Test Specificity Mouse CD90.2/Thy-1.2 alloantigen, Mr 18 kDa
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.