Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.2, Rat anti-Mouse, Biotin, Clone: 53-2.1, Novus Biologicals™
SDP

Catalog No. NB004395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004395 100 μg
NB004396 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB004395 Supplier Novus Biologicals Supplier No. NBP143609100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

CD90.2 Monoclonal specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD90.2
Applications Flow Cytometry
Classification Monoclonal
Clone 53-2.1
Concentration 5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry 1:10-1:1000
Formulation PBS (pH 7.2) with 0.1% gelatin
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Gene Symbols THY1
Host Species Rat
Immunogen The immunogen for this antibody was CD90.2.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7070
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.