Learn More
CD90 Mouse anti-Human, FITC, Clone: 5E10, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555595
Description
Reacts with a 25-35kDa molecule expressed on 1-4% of human fetal liver cells, cord blood cells, and bone marrow cells. Anti-CD90 reacts with a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+ CD4+ . The CD90+ CD34+ population is highly enriched for cells capable of long-term culture. Anti-CD90 is useful for enriching high proliferative potential colony-forming cells (HIPP-CFC) which are primative progenitor cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 5E10
Isotype: IgG1 κ
Species Reactivity: Hu
Flow Cytometry

Specifications
CD90 | |
Monoclonal | |
0.5mg/mL | |
THY1; Thy-1 antigen; Thy-1 membrane glycoprotein | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5.00E+10 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human HEL Cell Line | |
0.1 mg | |
Immunology | |
Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.