Learn More
CD90 Mouse anti-Human, FITC, Clone: 5E10, BD
Mouse Monoclonal Antibody
$128.60 - $279.50
Specifications
Antigen | CD90 |
---|---|
Clone | 5.00E+10 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The 5E10 monoclonal antibody specifically binds to human CD90 which is also known as Thy-1. CD90 is a 25-35 kDa glycophosphatidylinositol-anchored membrane glycoprotein of the Ig superfamily that is expressed on 1-4% of human fetal liver cells, cord blood cells, and bone marrow cells. The anti-CD90 antibody binds to a subset of immature CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is highly enriched for cells capable of long-term culture. The anti-CD90 antibody is useful for enriching high proliferative potential colony-forming cells (HIPP-CFC) that are primative progenitor cells.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD90 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey | |
Human HEL Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
5.00E+10 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
THY1; Thy-1 antigen; Thy-1 membrane glycoprotein |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.