Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90 Mouse anti-Human, Porcine, Rat, Clone: 2D7D11, Proteintech
SDP

Catalog No. 501737172 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7172 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7172 Supplier Proteintech Group Inc Supplier No. 667661IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

THY1 encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins. The encoded protein is involved in cell adhesion and cell communication in numerous cell types, but particularly in cells of the immune and nervous systems. The encoded protein is widely used as a marker for hematopoietic stem cells. This gene may function as a tumor suppressor in nasopharyngeal carcinoma. Alternative splicing results in multiple transcript variants.

Specifications

Antigen CD90
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D7D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene THY1
Gene Accession No. P01830, P04216
Gene Alias CD90, CDw90, FLJ33325, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1
Gene Symbols THY1
Host Species Mouse
Immunogen CD90 Fusion Protein Ag25603
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100271931, 24832, 7070
Target Species Human, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.