Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90 Mouse anti-Human, Porcine, Rat, Clone: 2D7D11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CD90 |
---|---|
Clone | 2D7D11 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7172
![]() ![]() |
Proteintech Group Inc
667661IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
THY1 encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins. The encoded protein is involved in cell adhesion and cell communication in numerous cell types, but particularly in cells of the immune and nervous systems. The encoded protein is widely used as a marker for hematopoietic stem cells. This gene may function as a tumor suppressor in nasopharyngeal carcinoma. Alternative splicing results in multiple transcript variants.Specifications
CD90 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD90, CDw90, FLJ33325, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1 | |
THY1 | |
IgG2a | |
Protein A | |
Antibody |
2D7D11 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig, Rat | |
P01830, P04216 | |
100271931, 24832, 7070 | |
CD90 Fusion Protein Ag25603 | |
Primary | |
-20°C | |
THY1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title