Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90/Thy1 Antibody (1aG4/C5), HRP, Novus Biologicals™
SDP

Catalog No. NB358809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358809 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358809 Supplier Novus Biologicals Supplier No. NBP144877H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Mouse, Human (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Functional Assay, Flow (Cell Surface), CyTOF

Specifications

Antigen CD90/Thy1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Functional Assay, Flow Cytometry (Cell Surface), CyTOF
Classification Monoclonal
Clone 1aG4/C5
Conjugate HRP
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Functional, Flow (Cell Surface), CyTOF-ready
Formulation PBS
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Host Species Mouse
Immunogen thymocytes from C3H/Di mice (alelic form: Thy-1.2 positive)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7070
Target Species Mouse, Human (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.