Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90/Thy1 Antibody (1aG4/C5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP44877646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP44877646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP44877646 Supplier Novus Biologicals Supplier No. NBP144877JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse, Human (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD90/Thy1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1aG4/C5
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Gene Symbols THY1
Host Species Mouse
Immunogen thymocytes from C3H/Di mice (alelic form: Thy-1.2 positive)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7070
Test Specificity The antibody 1aG4/C5 recognizes alloantigen Thy-1.2 of mouse Thy-1 (CD90); it does not cross-react with Thy-1.1 alloantigen. Thy-1 is a 18 kDa cell surface GPI linked glycoprotein (Immunoglobulin superfamily); in the mouse it is expressed on all thymocytes and peripheral T lymphocytes.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.