Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90/Thy1 Antibody (OX-7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10065543Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065543Y 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065543Y Supplier Novus Biologicals Supplier No. NB10065543AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen CD90/Thy1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone OX-7
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Gene Symbols THY1
Host Species Mouse
Immunogen Rat Thy 1 antigen
Purification Method Protein A purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7070
Test Specificity CD90
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.