Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Antibody (1032011), Biotin, Novus Biologicals™
SDP

Catalog No. NB175896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175896 Supplier Novus Biologicals Supplier No. FAB10581B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone 1032011
Conjugate Biotin
Formulation PBS
Gene Alias CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human CD94, Lys32-Ile179, Accession # Q13241.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3824
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.