Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (1H1), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | CD94 |
---|---|
Clone | 1H1 |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
CD94 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
KLRD1 | |
IgG1 | |
Unpurified |
1H1 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Innate Immunity | |
Ascites with 0.03% Sodium Azide | |
3824 | |
Purified recombinant fragment of human CD94 expressed in E. Coli. | |
Primary | |
21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title