Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Antibody (DX22) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064104 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064104 0.1 mg
NB017558 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064104 Supplier Novus Biologicals Supplier No. NB10064104
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone DX22
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100
Gene Accession No. Q13241
Gene Alias CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Gene Symbols KLRD1
Host Species Mouse
Immunogen Natural killer cell line
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3824
Test Specificity NB100-64104 is specific for human CD94, also known as KLRD1 (Killer cell lectin-like receptor, subfamily D, member 1). CD94 is expressed on natural killer (NK) cells and a subset of T lymphocytes. CD94 is found to associate with NKG2 to form a heterodimer which is involved in the inhibition of cell mediated cytotoxicity against cells bearing appropriate MHC class I allotypes. Clone DX22 is reported to inhibit the binding of CD94 to HLA-E(2) and HLA-G(4).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.