Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Antibody (MM0181-5F26), HRP, Novus Biologicals™
SDP

Catalog No. NB361292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361292 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361292 Supplier Novus Biologicals Supplier No. NBP212185H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry, CyTOF, Neutralization

Specifications

Antigen CD94
Applications Flow Cytometry, CyTOF, Neutralization
Classification Monoclonal
Clone MM0181-5F26
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready, Block/Neutralize
Formulation PBS
Gene Alias CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Host Species Mouse
Immunogen Human recombinant CD94
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3824
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.