Learn More
CD95 Mouse anti-Human, FITC, Clone: DX2, BD
Mouse Monoclonal Antibody
$164.70 - $332.00
Specifications
Antigen | CD95 |
---|---|
Clone | DX2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB556640
|
BD Biosciences
556640 |
50 Tests |
Each for $164.70
|
|
|||||
BDB555673
|
BD Biosciences
555673 |
100 Tests |
Each for $332.00
|
|
|||||
Description
Reacts with the 40-50kDa transmembrane human cell surface Fas or Apo-1 antigen. The Fas/Apo-1 antigen is a polypeptide which plays a role in the programmed sequence of events leading to cell death, termed apoptosis. DX2 also cross-reacts with with a subset of peripheral blood lymphocytes, monocytes and granulocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on lymphocytes is similar to that seen with human peripheral blood lymphocytes, being broadly expressed on all major lymphocyte subsets. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: DX2
Isotype: IgG1, κ
Cross Reactivity: Baboon, Cynomolgus
Flow Cytometry
Specifications
CD95 | |
Flow Cytometry | |
FITC | |
RUO | |
Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey | |
Human CD95-transfected L Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
DX2 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.