Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD96 Antibody (628211), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB347079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347079 Supplier Novus Biologicals Supplier No. FAB6199MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CD96
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 628211
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD96, Val22-Asp501, Accession # NP_005807
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10225
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.