Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD96 Antibody (6A6), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB153628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153628 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153628 Supplier Novus Biologicals Supplier No. NBP312037DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD96 Monoclonal antibody specifically detects CD96 in Mouse samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen CD96
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 6A6
Conjugate DyLight 594
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Host Species Rabbit
Immunogen This antibody was raised by immunising rats with murine CD96.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10225
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.