Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Antibody (2Q42), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP222000V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222000V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222000V Supplier Novus Biologicals Supplier No. NBP222000V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD97 Monoclonal specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD97
Applications Flow Cytometry
Classification Monoclonal
Clone 2Q42
Conjugate DyLight 405
Dilution Flow Cytometry
Gene Alias CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Gene Symbols ADGRE5
Host Species Rat
Immunogen Mouse recombinant CD97
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.