Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Antibody (MEM-180), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB500425W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500425W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500425W Supplier Novus Biologicals Supplier No. NB500425AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD97 Monoclonal specifically detects CD97 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD97
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-180
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Gene Alias CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Gene Symbols ADGRE5
Host Species Mouse
Immunogen PHA-activated peripheral blood cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 976
Test Specificity The antibody MEM-180 recognizes an unique epitope on CD97, a 75-85 kDa surface glycoprotein of G-protein-coupled receptor family, expressed on activated B and T lymphocytes, monocytes/macrophages, dendritic cells and granulocytes. HLDA VI; WS Code BP 415 HLDA VI; WS Code NL N-L023
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.