Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB323810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323810 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323810 Supplier Novus Biologicals Supplier No. NBP299764MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD97 Polyclonal antibody specifically detects CD97 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen CD97
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human CD97 (Accession#: NP_001775.2; Met 1-Gln 398)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.