Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224552 0.1 mg
NBP224552SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224552 Supplier Novus Biologicals Supplier No. NBP224552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

CD97 Polyclonal specifically detects CD97 in Human, Mouse, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD97
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml, Simple Western 1:10, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Alias CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Gene Symbols ADGRE5
Host Species Rabbit
Immunogen A portion of amino acids 220-270 of human Leukocyte Antigen CD97 was used as the immunogen
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 976
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_d)rat,mouse, opossum, chimpanzee and rhesus monkey.
Target Species Human, Mouse, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.