Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Antibody (SR1972), Novus Biologicals™
SDP

Catalog No. NB387916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387916 100 μg
NB387915 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387916 Supplier Novus Biologicals Supplier No. NBP322085100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD97 Monoclonal antibody specifically detects CD97 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen CD97
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SR1972
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CD97 (Uniprot #: P48960)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.