Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Isoform 1 Antibody (380903), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348906 Supplier Novus Biologicals Supplier No. FAB2529MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Human samples. It is validated for CyTOF, Flow Cytometry, Neutralization, Western Blot.

Specifications

Antigen CD97 Isoform 1
Applications CyTOF, Flow Cytometry, Neutralization, Western Blot
Classification Monoclonal
Clone 380903
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD97, TM7LN1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD97, Gln21-Gln398, Accession # NP_001775.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.