Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Isoform 1 Antibody (587702), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB173873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173873 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173873 Supplier Novus Biologicals Supplier No. FAB3734D
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD97 Isoform 1 Monoclonal antibody specifically detects CD97 Isoform 1 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CD97 Isoform 1
Applications Flow Cytometry
Classification Monoclonal
Clone 587702
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CD97, TM7LN1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD97v2, Gln24-His384, Accession # AAH06676
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.